Merck
Low-dose lithium uptake promotes longevity in humans and metazoans.
European journal of nutrition (2011-02-09)
Kim Zarse, Takeshi Terao, Jing Tian, Noboru Iwata, Nobuyoshi Ishii, Michael Ristow
ABSTRACT
Lithium is a nutritionally essential trace element predominantly contained in vegetables, plant-derived foods, and drinking water. Environmental lithium exposure and concurrent nutritional intake vary considerably in different regions. We here have analyzed the possibility that low-dose lithium exposure may affect mortality in both metazoans and mammals. Based on a large Japanese observational cohort, we have used weighted regression analysis to identify putative effects of tap water-derived lithium uptake on overall mortality. Independently, we have exposed Caenorhabditis elegans, a small roundworm commonly used for anti-aging studies, to comparable concentrations of lithium, and have quantified mortality during this intervention. In humans, we find here an inverse correlation between drinking water lithium concentrations and all-cause mortality in 18 neighboring Japanese municipalities with a total of 1,206,174 individuals (β = -0.661, p = 0.003). Consistently, we find that exposure to a comparably low concentration of lithium chloride extends life span of C. elegans (p = 0.047). Taken together, these findings indicate that long-term low-dose exposure to lithium may exert anti-aging capabilities and unambiguously decreases mortality in evolutionary distinct species.
Sigma-Aldrich
Lithium chloride, ReagentPlus®, 99%
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium chloride, for molecular biology, ≥99%
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Lithium chloride, ACS reagent, ≥99%
Sigma-Aldrich
Lithium chloride, BioUltra, for molecular biology, anhydrous, ≥99.0% (AT)