Merck
  • A high-throughput pharmaceutical screen identifies compounds with specific toxicity against BRCA2-deficient tumors.
A high-throughput pharmaceutical screen identifies compounds with specific toxicity against BRCA2-deficient tumors.
Clinical cancer research : an official journal of the American Association for Cancer Research (2009-12-17)
Bastiaan Evers, Eva Schut, Eline van der Burg, Tanya M Braumuller, David A Egan, Henne Holstege, Pauline Edser, David J Adams, Richard Wade-Martins, Peter Bouwman, Jos Jonkers
ABSTRACT
Hereditary breast cancer is partly explained by germline mutations in BRCA1 and BRCA2. Although patients carry heterozygous mutations, their tumors have typically lost the remaining wild-type allele. Selectively targeting BRCA deficiency may therefore constitute an important therapeutic approach. Clinical trials applying this principle are underway, but it is unknown whether the compounds tested are optimal. It is therefore important to identify alternative compounds that specifically target BRCA deficiency and to test new combination therapies to establish optimal treatment strategies. We did a high-throughput pharmaceutical screen on BRCA2-deficient mouse mammary tumor cells and isogenic controls with restored BRCA2 function. Subsequently, we validated positive hits in vitro and in vivo using mice carrying BRCA2-deficient mammary tumors. Three alkylators-chlorambucil, melphalan, and nimustine-displayed strong and specific toxicity against BRCA2-deficient cells. In vivo, these showed heterogeneous but generally strong BRCA2-deficient antitumor activity, with melphalan and nimustine doing better than cisplatin and the poly-(ADP-ribose)-polymerase inhibitor olaparib (AZD2281) in this small study. In vitro drug combination experiments showed synergistic interactions between the alkylators and olaparib. Tumor intervention studies combining nimustine and olaparib resulted in recurrence-free survival exceeding 330 days in 3 of 5 animals tested. We generated and validated a platform for identification of compounds with specific activity against BRCA2-deficient cells that translates well to the preclinical setting. Our data call for the re-evaluation of alkylators, especially melphalan and nimustine, alone or in combination with the poly-(ADP-ribose)-polymerase inhibitors, for the treatment of breast cancers with a defective BRCA pathway.
Sigma-Aldrich
LOPAC®1280 (International Version)
Sigma-Aldrich
Insulin human, meets USP testing specifications
Sigma-Aldrich
Insulin human, ≥95% (HPLC), semisynthetic, powder, non-sterile
Sigma-Aldrich
Insulin human, recombinant, expressed in yeast (proprietary host)
Sigma-Aldrich
Chlorambucil
Sigma-Aldrich
LOPAC®1280 - Small Scale, International Version
Sigma-Aldrich
Insulin human, recombinant, expressed in yeast, γ-irradiated, suitable for cell culture