Merck
  • Inhibition of BRD4 Attenuates ER Stress-induced Renal Ischemic-Reperfusion Injury.
Inhibition of BRD4 Attenuates ER Stress-induced Renal Ischemic-Reperfusion Injury.
International journal of biological sciences (2024-03-14)
Paula Diaz-Bulnes, Ramon M Rodríguez, Elisenda Banon-Maneus, María Laura Saiz, Cristian Ruiz Bernet, Viviana Corte-Iglesias, Maria Jose Ramirez-Bajo, Marta Lazo-Rodriguez, Isaac Tamargo-Gómez, Raúl R Rodrigues-Diez, Ana B Sanz, Carmen Diaz-Corte, Marta Ruiz-Ortega, Fritz Diekmann, Ana M Aransay, Carlos Lopez-Larrea, Beatriz Suarez-Alvarez
ABSTRACT
Renal ischemia-reperfusion injury (IRI) leads to endoplasmic reticulum (ER) stress, thereby initiating the unfolded protein response (UPR). When sustained, this response may trigger the inflammation and tubular cell death that acts to aggravate the damage. Here, we show that knockdown of the BET epigenetic reader BRD4 reduces the expression of ATF4 and XBP1 transcription factors under ER stress activation. BRD4 is recruited to the promoter of these highly acetylated genes, initiating gene transcription. Administration of the BET protein inhibitor, JQ1, one hour after renal damage induced by bilateral IRI, reveals reduced expression of ATF4 and XBP1 genes, low KIM-1 and NGAL levels and recovery of the serum creatinine and blood urea nitrogen levels. To determine the molecular pathways regulated by ATF4 and XBP1, we performed stable knockout of both transcription factors using CRISPR-Cas9 and RNA sequencing. The pathways triggered under ER stress were mainly XBP1-dependent, associated with an adaptive UPR, and partially regulated by JQ1. Meanwhile, treatment with JQ1 downmodulated most of the pathways regulated by ATF4 and related to the pathological processes during exacerbated UPR activation. Thus, BRD4 inhibition could be useful for curbing the maladaptive UPR activation mechanisms, thereby ameliorating the progression of renal disease.
Sigma-Aldrich
Anti-phospho RNA Pol II (Ser2), clone 3E7C7 Antibody, clone 3E7C7, from rat
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, 1 mg/mL, Upstate®
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit