Merck
Nrf2 Plays a Key Role in Erythropoiesis during Aging.
Antioxidants (Basel, Switzerland) (2024-04-27)
Serge Cedrick Toya Mbiandjeu, Angela Siciliano, Alessandro Mattè, Enrica Federti, Massimiliano Perduca, Davide Melisi, Immacolata Andolfo, Angela Amoresano, Achille Iolascon, Maria Teresa Valenti, Francesco Turrini, Michele Bovi, Arianna Pisani, Antonio Recchiuti, Domenico Mattoscio, Veronica Riccardi, Luca Dalle Carbonare, Carlo Brugnara, Narla Mohandas, Lucia De Franceschi
ABSTRACT
Aging is characterized by increased oxidation and reduced efficiency of cytoprotective mechanisms. Nuclear factor erythroid-2-related factor (Nrf2) is a key transcription factor, controlling the expression of multiple antioxidant proteins. Here, we show that Nrf2-/- mice displayed an age-dependent anemia, due to the combined contributions of reduced red cell lifespan and ineffective erythropoiesis, suggesting a role of Nrf2 in erythroid biology during aging. Mechanistically, we found that the expression of antioxidants during aging is mediated by activation of Nrf2 function by peroxiredoxin-2. The absence of Nrf2 resulted in persistent oxidation and overactivation of adaptive systems such as the unfolded protein response (UPR) system and autophagy in Nrf2-/- mouse erythroblasts. As Nrf2 is involved in the expression of autophagy-related proteins such as autophagy-related protein (Atg) 4-5 and p62, we found impairment of late phase of autophagy in Nrf2-/- mouse erythroblasts. The overactivation of the UPR system and impaired autophagy drove apoptosis of Nrf2-/- mouse erythroblasts via caspase-3 activation. As a proof of concept for the role of oxidation, we treated Nrf2-/- mice with astaxanthin, an antioxidant, in the form of poly (lactic-co-glycolic acid) (PLGA)-loaded nanoparticles (ATS-NPs) to improve its bioavailability. ATS-NPs ameliorated the age-dependent anemia and decreased ineffective erythropoiesis in Nrf2-/- mice. In summary, we propose that Nrf2 plays a key role in limiting age-related oxidation, ensuring erythroid maturation and growth during aging.
Sigma-Aldrich
Monoclonal Anti-CD44-FITC antibody produced in mouse, clone MEM-85, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-ACTIN antibody produced in rabbit, affinity isolated antibody