Merck
  • Potential Contribution of Cell Adhesion Molecule 1 to the Binding of SARS-CoV-2 Spike Protein to Mouse Nasal Mucosa.
Potential Contribution of Cell Adhesion Molecule 1 to the Binding of SARS-CoV-2 Spike Protein to Mouse Nasal Mucosa.
Cells, tissues, organs (2023-10-31)
Fuka Takeuchi, Aki Sugano, Azusa Yoneshige, Man Hagiyama, Takao Inoue, Akihiro Wada, Yutaka Takaoka, Akihiko Ito
ABSTRACT
Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) first infects the host nasal mucosa, where the viral spike protein binds to angiotensin-converting enzyme 2 (ACE2) on the mucosal cells. This study aimed at searching host cell surface molecules that could contribute to the infection in two views; abundance on host cells and affinity to the spike protein. Since the nasal mucosa is lined by respiratory and olfactory epithelia, and both express an immunoglobulin superfamily member cell adhesion molecule 1 (CADM1), whether CADM1 would participate in the spike protein binding was examined. Immunohistochemistry on the mouse nasal cavity detected CADM1 strongly in the olfactory epithelium at cell-cell contacts and on the apical surface but just faintly in the respiratory epithelium. In contrast, ACE2 was detected in the respiratory, not olfactory, epithelium. When mice were administered intranasally with SARS-CoV-2 S1 spike protein and an anti-CADM1 ectodomain antibody separately, both were detected exclusively on the olfactory, not respiratory, epithelium. Then, the antibody and S1 spike protein were administered intranasally to mice in this order with an interval of 1 h. After 3 h, S1 spike protein was detected as a protein aggregate floating in the nasal cavity. Next, S1 spike protein labeled with fluorescein was added to the monolayer cultures of epithelial cells exogenously expressing ACE2 or CADM1. Quantitative detection of fluorescein bound to the cells revealed that S1 spike protein bound to CADM1 with affinity half as high as to ACE2. Consistently, docking simulation analyses revealed that S1 spike protein could bind to CADM1 three-quarters as strongly as to ACE2 and that the interface of ACE2 was similar in both binding modes. Collectively, intranasal S1 spike protein appeared to prefer to accumulate on the olfactory epithelium, and CADM1 was suggested to contribute to this preference of S1 spike protein based on the molecular abundance and affinity.
Sigma-Aldrich
Anti-SynCAM antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution