Merck
  • PKM2 diverts glycolytic flux in dependence on mitochondrial one-carbon cycle.
PKM2 diverts glycolytic flux in dependence on mitochondrial one-carbon cycle.
Cell reports (2024-02-29)
Mohaned Benzarti, Laura Neises, Anais Oudin, Christina Krötz, Elodie Viry, Ernesto Gargiulo, Coralie Pulido, Maryse Schmoetten, Vitaly Pozdeev, Nadia I Lorenz, Michael W Ronellenfitsch, David Sumpton, Marc Warmoes, Christian Jaeger, Antoine Lesur, Björn Becker, Etienne Moussay, Jerome Paggetti, Simone P Niclou, Elisabeth Letellier, Johannes Meiser
ABSTRACT
Modeling tumor metabolism in vitro remains challenging. Here, we used galactose as an in vitro tool compound to mimic glycolytic limitation. In contrast to the established idea that high glycolytic flux reduces pyruvate kinase isozyme M2 (PKM2) activity to support anabolic processes, we have discovered that glycolytic limitation also affects PKM2 activity. Surprisingly, despite limited carbon availability and energetic stress, cells induce a near-complete block of PKM2 to divert carbons toward serine metabolism. Simultaneously, TCA cycle flux is sustained, and oxygen consumption is increased, supported by glutamine. Glutamine not only supports TCA cycle flux but also serine synthesis via distinct mechanisms that are directed through PKM2 inhibition. Finally, deleting mitochondrial one-carbon (1C) cycle reversed the PKM2 block, suggesting a potential formate-dependent crosstalk that coordinates mitochondrial 1C flux and cytosolic glycolysis to support cell survival and proliferation during nutrient-scarce conditions.
Sigma-Aldrich
Anti-SHMT2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
Sigma-Aldrich
Anti-PHGDH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Sigma-Aldrich
Anti-PSAT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody
Sigma-Aldrich
Anti-PSPH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution