Merck
  • PCDH12 loss results in premature neuronal differentiation and impeded migration in a cortical organoid model.
PCDH12 loss results in premature neuronal differentiation and impeded migration in a cortical organoid model.
Cell reports (2023-07-22)
Jennifer Rakotomamonjy, Lauren Rylaarsdam, Lucas Fares-Taie, Sean McDermott, Devin Davies, George Yang, Fikayo Fagbemi, Maya Epstein, Martín Fairbanks-Santana, Jean-Michel Rozet, Alicia Guemez-Gamboa
ABSTRACT
Protocadherins (PCDHs) are cell adhesion molecules that regulate many essential neurodevelopmental processes related to neuronal maturation, dendritic arbor formation, axon pathfinding, and synaptic plasticity. Biallelic loss-of-function variants in PCDH12 are associated with several neurodevelopmental disorders (NDDs). Despite the highly deleterious outcome resulting from loss of PCDH12, little is known about its role during brain development and disease. Here, we show that PCDH12 loss severely impairs cerebral organoid development, with reduced proliferative areas and disrupted laminar organization. 2D models further show that neural progenitor cells lacking PCDH12 prematurely exit the cell cycle and differentiate earlier when compared with wild type. Furthermore, we show that PCDH12 regulates neuronal migration and suggest that this could be through a mechanism requiring ADAM10-mediated ectodomain shedding and/or membrane recruitment of cytoskeleton regulators. Our results demonstrate a critical involvement of PCDH12 in cortical organoid development, suggesting a potential cause for the pathogenic mechanisms underlying PCDH12-related NDDs.
Sigma-Aldrich
Anti-WAVE-1 Antibody, clone K91/36, clone K91/36, from mouse
Sigma-Aldrich
Anti-MAP2 Antibody, clone AP20, clone AP20, Chemicon®, from mouse
Sigma-Aldrich
Anti-Sox2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture