Merck
Rational design of minimal hypoxia-inducible enhancers.
Biochemical and biophysical research communications (2008-04-12)
Stefan Kaluz, Milota Kaluzová, Eric J Stanbridge
ABSTRACT
The hypoxia-inducible factor (HIF) activates transcription via binding to the highly variable hypoxia-responsive elements (HREs). All hypoxia-inducible constructs described to date utilize multimers of naturally occurring HREs. Here, we describe the rational design of minimal hypoxia-inducible enhancers, conceptually equivalent to using an optimized HIF-binding site (HBS) as the building block. Optimizations of the HBS, spacing between HBSs, the distance from the minimal promoter, and orientation of HBSs allowed us to design constructs with high hypoxic activity. Activation of the 4xopt HBS (36bp) construct by hypoxia or HIF-1alpha and HIF-2alpha was comparable with that of the 4xEPO HRE (208bp) construct. The strong synergism between the properly arranged optimized HBSs was due to stimulation of high affinity HIF binding. Our data prove, for the first time, that it is possible to assemble artificial hypoxia-inducible enhancers from a single type of regulatory element-optimized HBS.
Product Description
Supelco
Streptavidin−Peroxidase Polymer, Ultrasensitive