Merck
  • Quantification of biofilm biomass by staining: Non-toxic safranin can replace the popular crystal violet.
Quantification of biofilm biomass by staining: Non-toxic safranin can replace the popular crystal violet.
Journal of microbiological methods (2017-08-15)
Pernille Ommen, Natalia Zobek, Rikke Louise Meyer
ABSTRACT
Crystal violet staining is commonly used for quantification of biofilm formation, although it is highly toxic. Here we test safranin as a non-toxic replacement. Safranin staining provided similar results as crystal violet, but with higher reproducibility. We therefore recommend safranin staining for biofilm biomass quantification.