Merck
  • Protocol to generate cardiac pericytes from human induced pluripotent stem cells.
Protocol to generate cardiac pericytes from human induced pluripotent stem cells.
STAR protocols (2023-04-29)
Mengcheng Shen, Shane Rui Zhao, Yaser Khokhar, Li Li, Yang Zhou, Chun Liu, Joseph C Wu
ABSTRACT
Cardiac pericytes are a critical yet enigmatic cell type within the coronary microvasculature. Since primary human cardiac pericytes are not readily accessible, we present a protocol to generate them from human induced pluripotent stem cells (hiPSCs). Our protocol involves several steps, including the generation of intermediate cell types such as mid-primitive streak, lateral plate mesoderm, splanchnic mesoderm, septum transversum, and epicardium, before deriving cardiac pericytes. With hiPSC-derived cardiac pericytes, researchers can decipher the mechanisms underlying coronary microvascular dysfunction. For complete details on the use and execution of this protocol, please refer to Shen et al.1.
Sigma-Aldrich
Monoclonal Anti-Calponin antibody produced in mouse, clone hCP, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Chondroitin Sulfate Proteoglycan Antibody, clone 9.2.27, clone 9.2.27, Chemicon®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Pricing and availability is not currently available.