Merck
  • SPTSSA variants alter sphingolipid synthesis and cause a complex hereditary spastic paraplegia.
SPTSSA variants alter sphingolipid synthesis and cause a complex hereditary spastic paraplegia.
Brain : a journal of neurology (2023-02-01)
Siddharth Srivastava, Hagar Mor Shaked, Kenneth Gable, Sita D Gupta, Xueyang Pan, Niranjanakumari Somashekarappa, Gongshe Han, Payam Mohassel, Marc Gotkine, Elizabeth Doney, Paula Goldenberg, Queenie K G Tan, Yi Gong, Benjamin Kleinstiver, Brian Wishart, Heidi Cope, Claudia Brito Pires, Hannah Stutzman, Rebecca C Spillmann, Reza Sadjadi, Orly Elpeleg, Chia-Hsueh Lee, Hugo J Bellen, Simon Edvardson, Florian Eichler, Teresa M Dunn
ABSTRACT
Sphingolipids are a diverse family of lipids with critical structural and signalling functions in the mammalian nervous system, where they are abundant in myelin membranes. Serine palmitoyltransferase, the enzyme that catalyses the rate-limiting reaction of sphingolipid synthesis, is composed of multiple subunits including an activating subunit, SPTSSA. Sphingolipids are both essential and cytotoxic and their synthesis must therefore be tightly regulated. Key to the homeostatic regulation are the ORMDL proteins that are bound to serine palmitoyltransferase and mediate feedback inhibition of enzymatic activity when sphingolipid levels become excessive. Exome sequencing identified potential disease-causing variants in SPTSSA in three children presenting with a complex form of hereditary spastic paraplegia. The effect of these variants on the catalytic activity and homeostatic regulation of serine palmitoyltransferase was investigated in human embryonic kidney cells, patient fibroblasts and Drosophila. Our results showed that two different pathogenic variants in SPTSSA caused a hereditary spastic paraplegia resulting in progressive motor disturbance with variable sensorineural hearing loss and language/cognitive dysfunction in three individuals. The variants in SPTSSA impaired the negative regulation of serine palmitoyltransferase by ORMDLs leading to excessive sphingolipid synthesis based on biochemical studies and in vivo studies in Drosophila. These findings support the pathogenicity of the SPTSSA variants and point to excessive sphingolipid synthesis due to impaired homeostatic regulation of serine palmitoyltransferase as responsible for defects in early brain development and function.
Sigma-Aldrich
Anti-ORMDL3 Antibody, from rabbit, purified by affinity chromatography