Merck
  • ΔNp63 drives dysplastic alveolar remodeling and restricts epithelial plasticity upon severe lung injury.
ΔNp63 drives dysplastic alveolar remodeling and restricts epithelial plasticity upon severe lung injury.
Cell reports (2022-12-15)
Aaron I Weiner, Gan Zhao, Hanna M Zayas, Nicolas P Holcomb, Stephanie Adams-Tzivelekidis, Joanna Wong, Maria E Gentile, Dyuthi Reddy, Joey Wei, Gargi Palashikar, Kwaku K Quansah, Andrew E Vaughan
ABSTRACT
The lung exhibits a robust, multifaceted regenerative response to severe injuries such as influenza infection, during which quiescent lung-resident epithelial progenitors participate in two distinct reparative pathways: functionally beneficial regeneration via alveolar type 2 (AT2) cell proliferation and differentiation, and dysplastic tissue remodeling via intrapulmonary airway-resident basal p63+ progenitors. Here we show that the basal cell transcription factor ΔNp63 is required for intrapulmonary basal progenitors to participate in dysplastic alveolar remodeling following injury. We find that ΔNp63 restricts the plasticity of intrapulmonary basal progenitors by maintaining either active or repressive histone modifications at key differentiation gene loci. Following loss of ΔNp63, intrapulmonary basal progenitors are capable of either airway or alveolar differentiation depending on their surrounding environment both in vitro and in vivo. Uncovering these regulatory mechanisms of dysplastic repair and lung basal cell fate choice highlight potential therapeutic targets to promote functional alveolar regeneration following severe lung injuries.
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Anti-acetyl-alpha tubulin Antibody, clone 6-11B-1, clone 6-11B-1, from mouse
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
A 83-01, ≥98% (HPLC)
Sigma-Aldrich
Donkey serum