Merck
  • Selective autophagy of RIPosomes maintains innate immune homeostasis during bacterial infection.
Selective autophagy of RIPosomes maintains innate immune homeostasis during bacterial infection.
The EMBO journal (2022-10-13)
Subhash Mehto, Kautilya Kumar Jena, Rina Yadav, Swatismita Priyadarsini, Pallavi Samal, Sivaram Krishna, Kollori Dhar, Ashish Jain, Nishant Ranjan Chauhan, Krushna C Murmu, Ramyasingh Bal, Rinku Sahu, Pundrik Jaiswal, Bhabani Sankar Sahoo, Srinivas Patnaik, Thomas A Kufer, Tor Erik Rusten, Swati Chauhan, Punit Prasad, Santosh Chauhan
ABSTRACT
The NOD1/2-RIPK2 is a key cytosolic signaling complex that activates NF-κB pro-inflammatory response against invading pathogens. However, uncontrolled NF-κB signaling can cause tissue damage leading to chronic diseases. The mechanisms by which the NODs-RIPK2-NF-κB innate immune axis is activated and resolved remain poorly understood. Here, we demonstrate that bacterial infection induces the formation of endogenous RIPK2 oligomers (RIPosomes) that are self-assembling entities that coat the bacteria to induce NF-κB response. Next, we show that autophagy proteins IRGM and p62/SQSTM1 physically interact with NOD1/2, RIPK2 and RIPosomes to promote their selective autophagy and limit NF-κB activation. IRGM suppresses RIPK2-dependent pro-inflammatory programs induced by Shigella and Salmonella. Consistently, the therapeutic inhibition of RIPK2 ameliorates Shigella infection- and DSS-induced gut inflammation in Irgm1 KO mice. This study identifies a unique mechanism where the innate immune proteins and autophagy machinery are recruited together to the bacteria for defense as well as for maintaining immune homeostasis.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Ubiquitin E1 Inhibitor, PYR-41, The Ubiquitin E1 Inhibitor, PYR-41 controls the biological activity of Ubiquitin E1. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.
Sigma-Aldrich
Anti-NOD2 Antibody, clone 2D9, clone 2D9, from mouse