Merck
  • Probenecid slows disease progression in a murine model of autosomal dominant polycystic kidney disease.
Probenecid slows disease progression in a murine model of autosomal dominant polycystic kidney disease.
Physiological reports (2023-04-07)
Sergey N Arkhipov, D'Anna L Potter, Regina F Sultanova, Daria V Ilatovskaya, Peter C Harris, Tengis S Pavlov
ABSTRACT
Development of autosomal dominant polycystic kidney disease (ADPKD) involves renal epithelial cell abnormalities. Cystic fluid contains a high level of ATP that, among other effects, leads to a reduced reabsorption of electrolytes in cyst-lining cells, and thus results in cystic fluid accumulation. Earlier, we demonstrated that Pkd1RC/RC mice, a hypomorphic model of ADPKD, exhibit increased expression of pannexin-1, a membrane channel capable of ATP release. In the current study, we found that human ADPKD cystic epithelia have higher pannexin-1 abundance than normal collecting ducts. We hypothesized that inhibition of pannexin-1 function with probenecid can be used to attenuate ADPKD development. Renal function in male and female Pkd1RC/RC and control mice was monitored between 9 and 20 months of age. To test the therapeutic effects of probenecid (a uricosuric agent and a pannexin-1 blocker), osmotic minipumps were implanted in male and female Pkd1RC/RC mice, and probenecid or vehicle was administered for 42 days until 1 year of age. Probenecid treatment improved glomerular filtration rates and slowed renal cyst formation in male mice (as shown in histopathology). The mechanistic effects of probenecid on sodium reabsorption and fluid transport were tested on polarized mpkCCDcl4 cells subjected to short-circuit current measurements, and in 3D cysts grown in Matrigel. In the mpkCCDcl4 epithelial cell line, probenecid elicited higher ENaC currents and attenuated in vitro cyst formation, indicating lower sodium and less fluid retention in the cysts. Our studies open new avenues of research into targeting pannexin-1 in ADPKD pathology.
Sigma-Aldrich
Monoclonal Anti-PANX1, (C-terminal) antibody produced in mouse, clone 2E3, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
IgG2a, Kappa from murine myeloma, clone UPC 10, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-PANX1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution