Merck
  • Germinated brown rice protects against glutamate toxicity in HT22 hippocampal neurons through the jnk-mediated apoptotic pathway via the GABAA receptor.
Germinated brown rice protects against glutamate toxicity in HT22 hippocampal neurons through the jnk-mediated apoptotic pathway via the GABAA receptor.
IBRO neuroscience reports (2023-01-03)
Sukrit Promtang, Chairat Turbpaiboon, Eve Mon Oo, Ladawan Khowawisetsut, Panapat Uawithya, Supin Chompoopong
ABSTRACT
The anti-apoptosis effect of germinated brown rice (GBR) focusing on differentiated HT22 cells results in improved nutritional values after the germination process of GBR which contains total phenolic compounds and γ-aminobutyric acid (GABA). Cell death induced by 5 mM glutamate was investigated for 24 h to determine whether GBR mediates cell death through GABA receptors by using antagonists. The results showed that GBR (100 µg/ml) suppressed glutamate-induced cytotoxicity and caused arrest at the G1/S phase of the cell cycle in differentiated HT22 cells. Furthermore, GBR significantly decreased the expression level of c-Jun, while its active form, p-c-Jun, is the downstream product of the JNK-mediated apoptotic pathway and causes subsequent cell death. In addition, bicuculline (12.5 nM), a GABAA antagonist, could eliminate GBR effects, but phaclofen (1 mM), a GABAB antagonist, could not. Surprisingly, GBR exhibited a better neuroprotective effect than a pure commercial GABA compound (0.115 µM). These results indicated that GBR possessed high anti-apoptotic activity and inhibited cell death in differentiated HT22 cells by perturbing re-entry of the cell cycle and apoptosis via the GABAA receptor. Hence, GBR could be further used as a valuable nutritional compound to prevent apoptosis-induced neurodegenerative diseases.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
HT-22 Mouse Hippocampal Neuronal Cell Line, HT-22 mouse neuronal cell line is a valuable cell model for studies of glutamate-induced toxicity in neuronal cells.
Sigma-Aldrich
Anti-phospho-c-Jun (Ser73) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-c-Jun Antibody, clone 6E4.4, clone 6E4.4, from mouse