Merck
  • Reduction of Pertussis Inflammatory Pathology by Therapeutic Treatment With Sphingosine-1-Phosphate Receptor Ligands by a Pertussis Toxin-Insensitive Mechanism.
Reduction of Pertussis Inflammatory Pathology by Therapeutic Treatment With Sphingosine-1-Phosphate Receptor Ligands by a Pertussis Toxin-Insensitive Mechanism.
The Journal of infectious diseases (2016-11-07)
Ciaran Skerry, Karen Scanlon, Jeremy Ardanuy, Drew Roberts, Li Zhang, Hugh Rosen, Nicholas H Carbonetti
ABSTRACT
Recent data have demonstrated the potential of sphingosine 1-phosphate (S1P) receptor (S1PR) agonism in the treatment of infectious diseases. A previous study used a murine model of Bordetella pertussis infection to demonstrate that treatment with the S1PR agonist AAL-R reduces pulmonary inflammation during infection. In the current study, we showed that this effect is mediated via the S1PR1 on LysM+ (myeloid) cells. Signaling via this receptor results in reduced lung inflammation and cellular recruitment as well as reduced morbidity and mortality in a neonatal mouse model of disease. Despite the fact that S1PRs are pertussis toxin-sensitive G protein-coupled receptors, the effects of AAL-R were pertussis toxin insensitive in our model. Furthermore, our data demonstrate that S1PR agonist administration may be effective at therapeutic time points. These results indicate a role for S1P signaling in B. pertussis-mediated pathology and highlight the possibility of host-targeted therapy for pertussis.
Sigma-Aldrich
AAL-R, ≥98% (HPLC)