Merck
  • Juvenile hormone III skipped bisepoxide is widespread in true bugs (Hemiptera: Heteroptera).
Juvenile hormone III skipped bisepoxide is widespread in true bugs (Hemiptera: Heteroptera).
Royal Society open science (2021-05-12)
Keiji Matsumoto, Toyomi Kotaki, Hideharu Numata, Tetsuro Shinada, Shin G Goto
ABSTRACT
Juvenile hormone (JH) plays important roles in almost every aspect of insect development and reproduction. JHs are a group of acyclic sesquiterpenoids, and their farnesol backbone has been chemically modified to generate a homologous series of hormones in some insect lineages. JH III (methyl farnesoate, 10,11-epoxide) is the most common JH in insects, but Lepidoptera (butterflies and moths) and 'higher' Diptera (suborder: Brachycera; flies) have developed their own unique JHs. Although JH was first proposed in the hemipteran suborder Heteroptera (true bugs), the chemical identity of the heteropteran JH was only recently determined. Furthermore, recent studies revealed the presence of a novel JH, JH III skipped bisepoxide (JHSB3), in some heteropterans, but its taxonomic distribution remains largely unknown. In the present study, we investigated JHSB3 production in 31 heteropteran species, covering almost all heteropteran lineages, through ultra-performance liquid chromatography coupled with tandem mass spectrometry. We found that all of the focal species produced JHSB3, indicating that JHSB3 is widespread in heteropteran bugs and the evolutionary occurrence of JHSB3 ascends to the common ancestor of Heteroptera.
Sigma-Aldrich
Juvenile hormone III, ≥65%, liquid, non-sterile