Merck
  • Opening KATP channels induces inflammatory tolerance and prevents chronic pain.
Opening KATP channels induces inflammatory tolerance and prevents chronic pain.
Brain, behavior, and immunity (2022-10-06)
Cheng Qian, Yixin Fan, Lijuan Zong, Chen Miao, Lu-Lu Ji, Li Wan, Rumeng Jia, Xinmiao Qin, Yu Wang, Qi Wu, Xue-You Tao, Lanxiang Hao, Liang Hu, Wen-Tao Liu
ABSTRACT
Current treatments for chronic pain are unsatisfactory, therefore, new therapeutics are urgently needed. Our previous study indicated that KATP channel openers have analgesic effects, but the underlying mechanism has not been elucidated. We speculated that KATP channel openers might increase suppressor of cytokine signaling (SOCS)-3 expression to induce inflammatory tolerance and attenuate chronic pain. Postoperative pain was induced by plantar incision to establish a chronic pain model. Growth arrest-specific 6 (Gas6)-/- and Axl-/- mice were used for signaling studies. The microglia cell line BV-2 was cultured for the in vitro experiments. The KATP channel opener significantly attenuated incision-induced mechanical allodynia in mice associated with the upregulated expression of SOCS3. Opening KATP channels induced the expression of SOCS3 in the Gas6/Axl signaling pathway in microglia, inhibited incision-induced mechanical allodynia by activating the Gas6/Axl-SOCS3 signaling pathway, and induced inflammatory tolerance to relieve neuroinflammation and postoperative pain. We demonstrated that opening of the KATP channel opening activated Gas6/Axl/SOCS3 signaling to induce inflammatory tolerance and relieve chronic pain. We explored a new target for anti-inflammatory and analgesic effects by regulating the innate immune system and provided a theoretical basis for clinical preemptive analgesia.
Sigma-Aldrich
IL-6 from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
IL-1β mouse, recombinant, expressed in E. coli, untagged, >95% (SDS-PAGE)
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining