Merck
  • Oral Nicotine Induces Oxidative Stress and Inflammation but Does Not Subvert Tumor Suppressor and DNA Repair Responses in Mice.
Oral Nicotine Induces Oxidative Stress and Inflammation but Does Not Subvert Tumor Suppressor and DNA Repair Responses in Mice.
Indian journal of clinical biochemistry : IJCB (2021-07-06)
Angom Ranjana Devi, Mahuya Sengupta, Dipu Mani Barman, Yashmin Choudhury
ABSTRACT
Nicotine, responsible for the addictive properties of tobacco, is widely used in nicotine replacement therapy for tobacco use cessation. We investigated the time-dependent effect of treatment with nicotine on the tumor suppressor, DNA repair and immune responses. Swiss Albino mice (laca strain) of both sexes received nicotine dissolved at a dose of 100 µg/ml in 2% sucrose for 24 weeks, by oral gavage, while age- and gender-matched controls received only 2% sucrose for the same period. Nicotine-treated and control mice were sacrificed 6, 16 and 24 weeks post-treatment, and their tissues evaluated for alterations in histology, oxidative stress, TNF-α levels, nitric oxide (NO) and myeloperoxidase (MPO) release, tumor suppressor response and DNA repair response. Statistical significance of results was determined using Students' t test. The tissues of nicotine treated mice exhibited a large number of multinucleated and binucleated cells, enlarged nuclei and non-uniform distribution of cells, significant increase in expression of TNF-α gene and serum TNF-α, and time-dependent significant increase in lipid peroxidation, protein carbonylation, NO and MPO release when compared to age-and gender-matched controls. The mRNA expression of the tumor suppressor gene p53, its primary regulator Mdm2, and the DNA repair genes Brca2 and Ape1 were significantly elevated, but the corresponding protein levels remained largely unaltered. In conclusion, treatment with nicotine caused oxidative stress and inflammation which can cause widespread cellular damage from the very onset of treatment, without subverting the tumor suppressor and DNA repair responses.
Sigma-Aldrich
Anti-MDM2 antibody produced in rabbit, affinity isolated antibody