Merck
  • On-the-fly investigation of XUV excited large molecular ions using a high harmonic generation light source.
On-the-fly investigation of XUV excited large molecular ions using a high harmonic generation light source.
Scientific reports (2022-08-02)
Marius Hervé, Alexie Boyer, Richard Brédy, Abdul-Rahman Allouche, Isabelle Compagnon, Franck Lépine
ABSTRACT
We present experiments where extreme ultraviolet femtosecond light pulses are used to photoexcite large molecular ions at high internal energy. This is done by combining an electrospray ionization source and a mass spectrometer with a pulsed light source based on high harmonic generation. This allows one to study the interaction between high energy photons and mass selected ions in conditions that are accessible on large-scale facilities. We show that even without an ion trapping device, systems as large as a protein can be studied. We observe light induced dissociative ionization and proton migration in model systems such as reserpine, insulin and cytochrome c. These results offer new perspectives to perform time-resolved experiments with ultrashort pulses at the heart of the emerging field of attosecond chemistry.
Sigma-Aldrich
Insulin human, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis
Pricing and availability is not currently available.