Merck
  • Beta-glucuronidase-inhibitory activity and hepatoprotective effect of Ganoderma lucidum.
Beta-glucuronidase-inhibitory activity and hepatoprotective effect of Ganoderma lucidum.
Biological & pharmaceutical bulletin (1999-03-17)
D H Kim, S B Shim, N J Kim, I S Jang
ABSTRACT
To prove the relationship between the fluctuation in serum beta-glucuronidase level and hepatotoxicity, an inhibitor of beta-glucuronidase from G. lucidum was isolated and its hepatoprotective activity was investigated. The ether fraction of G. lucidum, which had potent beta-glucuronidase-inhibitory activity, protected against CCl4-induced liver injury. From this ether fraction, ganoderenic acid A, was isolated as the potent inhibitor of beta-glucuronidase. It had a potent hepatoprotective effect against CCl4-induced liver injury. These results suggest that the beta-glucuronidase seems to be closely related to liver injury, which could be prevented by beta-glucuronidase inhibitors.
Sigma-Aldrich
β-Glucuronidase from abalone, aqueous solution, β-glucuronidase ≥100,000 units/mL
Pricing and availability is not currently available.