Merck
Innate Lymphoid Cells in the Induction of Obesity.
Cell reports (2019-07-04)
Takaharu Sasaki, Kazuyo Moro, Tetsuya Kubota, Naoto Kubota, Tamotsu Kato, Hiroshi Ohno, Susumu Nakae, Hirohisa Saito, Shigeo Koyasu
ABSTRACT
Complex interactions between immune cells are an important component in the induction of obesity. Here, we show that Il2rg-/-Rag2-/- mice lacking all lymphocytes are resistant to diet-induced obesity. Transplantation of bone marrow cells from Rag2-/- mice, which lack only acquired immune cells, into Il2rg-/-Rag2-/- mice abolishes this resistance, indicating a role for innate lymphoid cells (ILCs) in this process. Mice lacking ILC2 or ILC3 cells, but not natural killer cells, are resistant to obesity. Adoptive transfer of naive ILC2s isolated from the small intestine (SI), but not ILC2s from white adipose tissue (WAT), restores the induction of diet-induced obesity in Il2rg-/-Rag2-/- mice. Analysis of transcriptional differences reveals that SI-ILC2s express higher levels of IL-2 than do WAT-ILC2s and that blockade of IL-2 signaling impairs weight gain and reduces the populations of ILC2s and ILC3s in the SI, suggesting a role for the IL-2/ILC2/3 axis in the induction of obesity.
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer