Merck
  • Sideroflexin 4 is a complex I assembly factor that interacts with the MCIA complex and is required for the assembly of the ND2 module.
Sideroflexin 4 is a complex I assembly factor that interacts with the MCIA complex and is required for the assembly of the ND2 module.
Proceedings of the National Academy of Sciences of the United States of America (2022-03-26)
Thomas D Jackson, Jordan J Crameri, Linden Muellner-Wong, Ann E Frazier, Catherine S Palmer, Luke E Formosa, Daniella H Hock, Kenji M Fujihara, Tegan Stait, Alice J Sharpe, David R Thorburn, Michael T Ryan, David A Stroud, Diana Stojanovski
ABSTRACT
SignificanceMitochondria are double-membraned eukaryotic organelles that house the proteins required for generation of ATP, the energy currency of cells. ATP generation within mitochondria is performed by five multisubunit complexes (complexes I to V), the assembly of which is an intricate process. Mutations in subunits of these complexes, or the suite of proteins that help them assemble, lead to a severe multisystem condition called mitochondrial disease. We show that SFXN4, a protein that causes mitochondrial disease when mutated, assists with the assembly of complex I. This finding explains why mutations in SFXN4 cause mitochondrial disease and is surprising because SFXN4 belongs to a family of amino acid transporter proteins, suggesting that it has undergone a dramatic shift in function through evolution.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Roche
ATP Bioluminescence Assay Kit CLS II, sufficient for 800 assays (tubes), kit of 1 (2 components), suitable for detection
Sigma-Aldrich
Anti-TIMMDC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution