Merck
CD226hiCD8+ T Cells Are a Prerequisite for Anti-TIGIT Immunotherapy.
Cancer immunology research (2020-04-09)
Hyung-Seung Jin, Minkyung Ko, Da-Som Choi, June Hyuck Kim, Dong-Hee Lee, Seong-Ho Kang, Inki Kim, Hee Jin Lee, Eun Kyung Choi, Kyu-Pyo Kim, Changhoon Yoo, Yoon Park
ABSTRACT
Clinical trials are evaluating the efficacy of anti-TIGIT for use as single-agent therapy or in combination with programmed death 1 (PD-1)/programmed death-ligand 1 blockade. How and whether a TIGIT blockade will synergize with immunotherapies is not clear. Here, we show that CD226loCD8+ T cells accumulate at the tumor site and have an exhausted phenotype with impaired functionality. In contrast, CD226hiCD8+ tumor-infiltrating T cells possess greater self-renewal capacity and responsiveness. Anti-TIGIT treatment selectively affects CD226hiCD8+ T cells by promoting CD226 phosphorylation at tyrosine 322. CD226 agonist antibody-mediated activation of CD226 augments the effect of TIGIT blockade on CD8+ T-cell responses. Finally, mFOLFIRINOX treatment, which increases CD226hiCD8+ T cells in patients with pancreatic ductal adenocarcinoma, potentiates the effects of TIGIT or PD-1 blockade. Our results implicate CD226 as a predictive biomarker for cancer immunotherapy and suggest that increasing numbers of CD226hiCD8+ T cells may improve responses to anti-TIGIT therapy.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-CD226/DNAM-1 Antibody, clone NewE1 (Azide Free), clone NewE1, from mouse