Merck
  • A transgenic mouse embryonic stem cell line for puromycin selection of V0V interneurons from heterogenous induced cultures.
A transgenic mouse embryonic stem cell line for puromycin selection of V0V interneurons from heterogenous induced cultures.
Stem cell research & therapy (2022-03-30)
Jennifer Pardieck, Manwal Harb, Shelly E Sakiyama-Elbert
ABSTRACT
Spinal interneurons (INs) relay sensory and motor control information between the brain and body. When this relay circuitry is disrupted from injury or disease, it is devastating to patients due to the lack of native recovery in central nervous system (CNS) tissues. Obtaining a purified population of INs is necessary to better understand their role in normal function and as potential therapies in CNS. The ventral V0 (V0V) INs are excitatory neurons involved in locomotor circuits and are thus of interest for understanding normal and pathological spinal cord function. To achieve scalable amounts of V0V INs, they can be derived from pluripotent sources, such as mouse embryonic stem cells (mESCs), but the resultant culture is heterogenous, obscuring the specific role of V0V INs. This study generated a transgenic mESC line to enrich V0V INs from induced cultures to allow for a scalable, enriched population for future in vitro and in vivo studies. The transgenic Evx1-PAC mESC line was created by CRISPR-Cas9-mediated insertion of puromycin-N-acetyltransferase (PAC) into the locus of V0V IN marker Evx1. Evx1 and PAC mRNA expression were measured by qPCR. Viability staining helped establish the selection protocol for V0V INs derived from Evx1-PAC mESCs inductions. Immunostaining was used to examine composition of selected inductions. Cultures were maintained up to 30 days to examine maturation by expression of mature/synaptic markers, determined by immunostaining, and functional activity in co-cultures with selected motor neurons (MNs) and V2a INs on microelectrode arrays (MEAs). V0V IN inductions were best selected with 4 µg/mL puromycin on day 10 to 11 and showed reduction of other IN populations and elimination of proliferative cells. Long-term selected cultures were highly neuronal, expressing neuronal nuclear marker NeuN, dendritic marker MAP2, pre-synaptic marker Bassoon, and glutamatergic marker VGLUT2, with some cholinergic VAChT-expressing cells. Functional studies on MEAs showed that co-cultures with MNs or MNs plus V2a INs created neuronal networks with synchronized bursting. Evx1-PAC mESCs can be used to purify V0V IN cultures for largely glutamatergic neurons that can be used in network formation studies or for rodent models requiring transplanted V0V INs.
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid
Sigma-Aldrich
Anti-Microtubule-Associated Protein 2 (MAP2) Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Sigma-Aldrich
Goat serum
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Accutase® solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Purmorphamine, InSolution, ≥98%, 50 mM in DMSO, induces osteoblast differentiation in C3H10T1/2 multipotent mesenchymal progenitor cells
Sigma-Aldrich
ESGRO® Recombinant Mouse LIF Protein, ESGRO Leukemia Inhibitory Factor (LIF) supplement for mouse ES cell culture. Each vial contains 10^6 units/ml
Sigma-Aldrich
Anti-VGluT2 Antibody, serum, from guinea pig
Sigma-Aldrich
Anti-Olig2 Antibody, clone 211F1.1, clone 211F1.1, from mouse