Merck
  • Circulating Lipocalin-2 level is positively associated with cognitive impairment in patients with metabolic syndrome.
Circulating Lipocalin-2 level is positively associated with cognitive impairment in patients with metabolic syndrome.
Scientific reports (2022-03-19)
Kanokporn Pinyopornpanish, Arintaya Phrommintikul, Chaisiri Angkurawaranon, Sirinart Kumfu, Salita Angkurawaranon, Uten Yarach, Nida Buawangpong, Nipon Chattipakorn, Siriporn C Chattipakorn
ABSTRACT
The association between Lipocalin-2 (LCN2) and cognition in patients with metabolic syndrome (MetS) has not been thoroughly investigated. We aimed to evaluate whether serum LCN2 levels are associated with the alteration of cognitive function in patients with MetS. The total of 191 non-demented participants with MetS were enrolled onto the study in 2015, and a cohort study was conducted in a subpopulation in 2020. After adjustment for sex, age, waist circumference, creatinine levels, and HbA1C, an association between the higher serum LCN2 levels and the lower Montreal cognitive assessment (MoCA) scores was observed (B = - 0.045; 95%CI - 0.087, - 0.004; p 0.030). A total of 30 participants were followed-up in 2020. Serum LCN2 levels were decreased in correlation with age (23.31 ± 12.32 ng/ml in 2015 and 15.98 ± 11.28 ng/ml in 2020, p 0.024), while other metabolic parameters were unchanged. Magnetic resonance imaging studies were conducted on a subsample of patients in 2020 (n = 15). Associations between high serum LCN2 levels from 2015 and 2020 and changes in brain volume of hippocampus and prefrontal cortex from 2020 have been observed. These findings suggest a relationship between changes of the level of circulating LCN2, cognitive impairment, and changes in brain volume in patients with MetS. However, further investigation is still needed to explore the direct effect of circulating LCN2 on the cognition of MetS patients.
Sigma-Aldrich
Human Lipocalin-2 / NGAL ELISA Kit, for serum, plasma, cell culture supernatant and urine