Merck
  • An Elaborate New Linker System Significantly Enhances the Efficacy of an HER2-Antibody-Drug Conjugate against Refractory HER2-Positive Cancers.
An Elaborate New Linker System Significantly Enhances the Efficacy of an HER2-Antibody-Drug Conjugate against Refractory HER2-Positive Cancers.
Advanced science (Weinheim, Baden-Wurttemberg, Germany) (2021-10-20)
Seol Hwa Shin, Yun-Hee Park, Seok Soon Park, Eun Jin Ju, Jin Park, Eun Jung Ko, Dong Jun Bae, Sang-Yeob Kim, Chul-Woong Chung, Ho Young Song, Se Jin Jang, Seong-Yun Jeong, Si Yeol Song, Eun Kyung Choi
ABSTRACT
Human epidermal growth factor receptor 2 (HER2) is overexpressed in breast and gastric cancers and this causes poor clinical outcomes. Although both T-DM1 and Enhertu are approved as an HER2-targeting antibody-drug conjugate (ADC), the effects of these drugs are still not satisfactory to eradicate diverse tumors expressing HER2. To address this shortfall in HER2-targeted therapeutics, an elaborate cleavable linker is created and a novel HER2-targeting ADC composed with trastuzumab and monomethyl auristatin F, which is being investigated in a phase 1 clinical trial and is referred to as LegoChem Bisciences-ADC (LCB-ADC). LCB-ADC displays a higher cytotoxic potency than T-DM1 and it also has a higher G2/M arrest ratio. In animal studies, LCB-ADC produces noticeable tumor growth inhibition compared with trastuzumab or T-DM1 in an HER2 high-expressing N87 xenograft tumor. Especially, LCB-ADC shows good efficacy in terms of suppressing tumor growth in a patient-derived xenograft (PDX) model of HER2-positive gastric cancer as well as in T-DM1-resistant models such as HER2 low-expressing HER2 low expressing JIMT-1 xenograft tumor and PDX. Collectively, the results demonstrate that LCB-ADC with the elaborate linker has a higher efficacy and greater biostability than its ADC counterparts and may successfully treat cancers that are nonresponsive to previous therapeutics.
Sigma-Aldrich
FPTase, Rat, Recombinant, E. coli, A heterodimeric enzyme that catalyzes the transfer of a 15-carbon isoprenoid group to a variety of cellular proteins including Ras.
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)