Merck
  • 4-Bromophenacyl bromide specifically inhibits rhoptry secretion during Toxoplasma invasion.
4-Bromophenacyl bromide specifically inhibits rhoptry secretion during Toxoplasma invasion.
PloS one (2009-12-04)
Sandeep Ravindran, Melissa B Lodoen, Steven H L Verhelst, Matthew Bogyo, John C Boothroyd
ABSTRACT
Toxoplasma gondii is a eukaryotic parasite of the phylum Apicomplexa that is able to infect a wide variety of host cells. During its active invasion process it secretes proteins from discrete secretory organelles: the micronemes, rhoptries and dense granules. Although a number of rhoptry proteins have been shown to be involved in important interactions with the host cell, very little is known about the mechanism of secretion of any Toxoplasma protein into the host cell. We used a chemical inhibitor of phospholipase A2s, 4-bromophenacyl bromide (4-BPB), to look at the role of such lipases in the secretion of Toxoplasma proteins. We found that 4-BPB was a potent inhibitor of rhoptry secretion in Toxoplasma invasion. This drug specifically blocked rhoptry secretion but not microneme secretion, thus effectively showing that the two processes can be de-coupled. It affected parasite motility and invasion, but not attachment or egress. Using propargyl- or azido-derivatives of the drug (so-called click chemistry derivatives) and a series of 4-BPB-resistant mutants, we found that the drug has a very large number of target proteins in the parasite that are involved in at least two key steps: invasion and intracellular growth. This potent compound, the modified "click-chemistry" forms of it, and the resistant mutants should serve as useful tools to further study the processes of Toxoplasma early invasion, in general, and rhoptry secretion, in particular.
Sigma-Aldrich
2,4′-Dibromoacetophenone, >98%
Supelco
2,4′-Dibromoacetophenone, for HPLC derivatization, LiChropur, ≥99.0% (HPLC)
Sigma-Aldrich
Cytochalasin D, Ready Made Solution, from Zygosporium mansonii, 5 mg/mL in DMSO
Sigma-Aldrich
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, 97%
Sigma-Aldrich
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder