Merck
  • A Simple Enzymatic Method for Production of a Wide Variety of D-Amino Acids Using L-Amino Acid Oxidase from Rhodococcus sp. AIU Z-35-1.
A Simple Enzymatic Method for Production of a Wide Variety of D-Amino Acids Using L-Amino Acid Oxidase from Rhodococcus sp. AIU Z-35-1.
Enzyme research (2010-11-05)
Kimiyasu Isobe, Hiroshi Tamauchi, Ken-Ichi Fuhshuku, Shouko Nagasawa, Yasuhisa Asano
ABSTRACT
A simple enzymatic method for production of a wide variety of D-amino acids was developed by kinetic resolution of DL-amino acids using L-amino acid oxidase (L-AAO) with broad substrate specificity from Rhodococcus sp. AIU Z-35-1. The optimum pH of the L-AAO reaction was classified into three groups depending on the L-amino acids as substrate, and their respective activities between pH 5.5 and 8.5 accounted for more than 60% of the optimum activity. The enzyme was stable in the range from pH 6.0 to 8.0, and approximately 80% of the enzyme activity remained after incubation at 40°C for 60 min at pH 7.0. D-Amino acids such as D-citrulline, D-glutamine, D-homoserine or D-arginine, which are not produced by D-aminoacylases or D-hydantoinases, were produced from the racemic mixture within a 24-hr reaction at 30°C and pH 7.0. Thus, the present method using L-AAO was versatile for production of a wide variety of D-amino acids.
Sigma-Aldrich
L-Homoserine