Merck
  • Lung emphysema and impaired macrophage elastase clearance in mucolipin 3 deficient mice.
Lung emphysema and impaired macrophage elastase clearance in mucolipin 3 deficient mice.
Nature communications (2022-01-16)
Barbara Spix, Elisabeth S Butz, Cheng-Chang Chen, Anna Scotto Rosato, Rachel Tang, Aicha Jeridi, Veronika Kudrina, Eva Plesch, Philipp Wartenberg, Elisabeth Arlt, Daria Briukhovetska, Meshal Ansari, Gizem Günes Günsel, Thomas M Conlon, Amanda Wyatt, Sandra Wetzel, Daniel Teupser, Lesca M Holdt, Fabien Ectors, Ingrid Boekhoff, Ulrich Boehm, Jaime García-Añoveros, Paul Saftig, Martin Giera, Sebastian Kobold, Herbert B Schiller, Susanna Zierler, Thomas Gudermann, Christian Wahl-Schott, Franz Bracher, Ali Önder Yildirim, Martin Biel, Christian Grimm
ABSTRACT
Lung emphysema and chronic bronchitis are the two most common causes of chronic obstructive pulmonary disease. Excess macrophage elastase MMP-12, which is predominantly secreted from alveolar macrophages, is known to mediate the development of lung injury and emphysema. Here, we discovered the endolysosomal cation channel mucolipin 3 (TRPML3) as a regulator of MMP-12 reuptake from broncho-alveolar fluid, driving in two independently generated Trpml3-/- mouse models enlarged lung injury, which is further exacerbated after elastase or tobacco smoke treatment. Mechanistically, using a Trpml3IRES-Cre/eR26-τGFP reporter mouse model, transcriptomics, and endolysosomal patch-clamp experiments, we show that in the lung TRPML3 is almost exclusively expressed in alveolar macrophages, where its loss leads to defects in early endosomal trafficking and endocytosis of MMP-12. Our findings suggest that TRPML3 represents a key regulator of MMP-12 clearance by alveolar macrophages and may serve as therapeutic target for emphysema and chronic obstructive pulmonary disease.
Sigma-Aldrich
holo-Transferrin human, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Latrunculin B from Latruncula magnifica, ≥80% (HPLC), solid
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
Anti-CD3, T Cell antibody produced in rabbit, whole antiserum
Roche
cOmplete, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks