Merck
  • Secondary Cerebellar Cortex Injury in Albino Male Rats after MCAO: A Histological and Biochemical Study.
Secondary Cerebellar Cortex Injury in Albino Male Rats after MCAO: A Histological and Biochemical Study.
Biomedicines (2021-09-29)
Aziza R Alrafiah
ABSTRACT
The present study focused on secondary injury following the middle cerebral artery (MCA) occlusion in rats not linked to the MCA's feeding zone. This entity has been very rarely studied. Additionally, this study investigated the rates of expression of five fundamental angiogenic biomarkers called endoglin, vascular endothelial growth factors-A (VEGF-A), endothelin-1 (ET-1), 2granulocyte colony-stimulating factor (G-CSF), and angiopoietin-using the MCA occlusion (MCAO) model. The random allocation of twelve adult male albino rats was in two groups. As a sham control group, six rats were used. This group was subjected to a sham operation without MCAO. The MCAO group consisted of six rats that were subjected to MCAO operation. After three days, the rats were sacrificed. The cerebellar specimens were immediately processed for light microscopic examination. An angiogenic biomarkers multiplex assay from multiplex was used to assess endoglin levels, VEGF-A, ET-1, angiopoietin-2, and G-CSF in serum samples. Hematoxylin and eosin-stained sections showed that the cerebellar cortex of rats of the MCAO group was more affected than the sham control group. Furthermore, Nissl stain and immunohistochemical analysis revealed an apparent increase in the number of positive immunoreactive in the cerebellar cortex and an evident decrease in Nissl granules in Purkinje cells of the MCAO rats, in contrast to the control rats. In addition, there was a significant increase in angiogenic factors VEGF-A, ET-1, angiopoietin-2, and endoglin. Interestingly, there was an increase in the G-CSF but a non-significant in the MCAO rats compared to the control rats. Furthermore, there was a significant correlation between the angiopoietin-2 and ET-1, and between G-CSF and ET-1. VEGF-A also exhibited significant positive correlations with the G-CSF serum level parameter, Endoglin, and ET-1. Rats subjected to MCAO are a suitable model to study secondary injury away from MCA's feeding zone. Additionally, valuable insights into the association and interaction between altered angiogenic factors and acute ischemic stroke induced by MCAO in rats.
Product Description
Millipore
MILLIPLEX® Mouse Angiogenesis/Growth Factor Magnetic Bead Panel - Cancer Multiplex Assay, for the simultaneous quantification of multiple analytes