Merck
  • Quality of the Haemophilus influenzae type b (Hib) antibody response induced by diphtheria-tetanus-acellular pertussis/Hib combination vaccines.
Quality of the Haemophilus influenzae type b (Hib) antibody response induced by diphtheria-tetanus-acellular pertussis/Hib combination vaccines.
Clinical and vaccine immunology : CVI (2007-08-19)
Philippe A Denoël, David Goldblatt, Isabel de Vleeschauwer, Jeanne-Marie Jacquet, Michael E Pichichero, Jan T Poolman
ABSTRACT
It has been repeatedly observed that mixing Haemophilus influenzae type b (Hib) conjugate vaccines with acellular pertussis-containing vaccines (diphtheria-tetanus-acellular pertussis [DTPa]) resulted in a reduced magnitude of the anti-polyriboseribitolphosphate antibody response compared to that obtained when Hib vaccines were administered separately and not mixed. Nevertheless, the quality and functionality of the immune responses have been shown to be the same. With the purpose of investigating the quality of the anti-Hib immune responses that are elicited under different vaccination regimens, we report here four primary and booster-based pediatric clinical trials in which Hib vaccine was either mixed with DTPa or diphtheria-tetanus-whole-cell pertussis (DTPw)-based vaccines or was coadministered. Our results show that avidity maturation of the antibodies was lower when primary vaccination involved DTPa mixed with Hib compared to when DTPa and Hib were coadministered. No such difference was observed between mixed and separately administered Hib when associated with DTPa-hepatitis B virus-inactivated poliovirus or DTPw-based vaccines. All different combinations and regimens elicited the same opsonophagocytic and bactericidal activity as well as the same ability to protect in a passive infant rat protection assay. The functional activity of mixed DTPa-based and Hib vaccines was similar to that of mixed DTPw-based/Hib combinations. In conclusion, in vitro and in vivo data as well as postmarketing vaccine effectiveness data attest to the ability of DTPa-based/Hib combination vaccines to effectively prevent Hib-induced disease in children.
Sigma-Aldrich
Monoclonal Anti-Human IgG2 (Fc specific) antibody produced in mouse, clone HP-6002, ascites fluid
Sigma-Aldrich
Anti-Human IgG (γ-chain specific), F(ab′)2 fragment−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution