Merck
INFOGEST static in vitro simulation of gastrointestinal food digestion.
Nature protocols (2019-03-20)
André Brodkorb, Lotti Egger, Marie Alminger, Paula Alvito, Ricardo Assunção, Simon Ballance, Torsten Bohn, Claire Bourlieu-Lacanal, Rachel Boutrou, Frédéric Carrière, Alfonso Clemente, Milena Corredig, Didier Dupont, Claire Dufour, Cathrina Edwards, Matt Golding, Sibel Karakaya, Bente Kirkhus, Steven Le Feunteun, Uri Lesmes, Adam Macierzanka, Alan R Mackie, Carla Martins, Sébastien Marze, David Julian McClements, Olivia Ménard, Mans Minekus, Reto Portmann, Cláudia N Santos, Isabelle Souchon, R Paul Singh, Gerd E Vegarud, Martin S J Wickham, Werner Weitschies, Isidra Recio
ABSTRACT
Developing a mechanistic understanding of the impact of food structure and composition on human health has increasingly involved simulating digestion in the upper gastrointestinal tract. These simulations have used a wide range of different conditions that often have very little physiological relevance, and this impedes the meaningful comparison of results. The standardized protocol presented here is based on an international consensus developed by the COST INFOGEST network. The method is designed to be used with standard laboratory equipment and requires limited experience to encourage a wide range of researchers to adopt it. It is a static digestion method that uses constant ratios of meal to digestive fluids and a constant pH for each step of digestion. This makes the method simple to use but not suitable for simulating digestion kinetics. Using this method, food samples are subjected to sequential oral, gastric and intestinal digestion while parameters such as electrolytes, enzymes, bile, dilution, pH and time of digestion are based on available physiological data. This amended and improved digestion method (INFOGEST 2.0) avoids challenges associated with the original method, such as the inclusion of the oral phase and the use of gastric lipase. The method can be used to assess the endpoints resulting from digestion of foods by analyzing the digestion products (e.g., peptides/amino acids, fatty acids, simple sugars) and evaluating the release of micronutrients from the food matrix. The whole protocol can be completed in ~7 d, including ~5 d required for the determination of enzyme activities.
Product Description
Millipore
Bile bovine, dried, unfractionated
Sigma-Aldrich
Methyltrioctylammonium hydrogen sulfate, ≥95.0% (T)
Sigma-Aldrich
Amyloglucosidase from Aspergillus niger, ammonium sulfate suspension, ≥40 units/mg protein