Merck
  • A chemoproteomic method for identifying cellular targets of covalent kinase inhibitors.
A chemoproteomic method for identifying cellular targets of covalent kinase inhibitors.
Genes & cancer (2016-08-24)
Ying-Chu Chen, Chao Zhang
ABSTRACT
Protein kinases are attractive drug targets for numerous human diseases including cancers, diabetes and neurodegeneration. A number of kinase inhibitors that covalently target a cysteine residue in their target kinases have recently entered use in the cancer clinic. Despite the advantages of covalent kinases inhibitors, their inherent reactivity can lead to non-specific binding to other cellular proteins and cause off- target effects in cells. It is thus essential to determine the identity of these off targets in order to fully account for the phenotype and to improve the selectivity and efficacy of covalent inhibitors. Herein we present a detailed protocol for a chemoproteomic method to enrich and identify cellular targets of covalent kinase inhibitors.
Sigma-Aldrich
α,α,α′,α′-Tetramethyl-1,3-benzenedipropionic acid, 97%
Millipore
Protein A Agarose, Protein A Agarose Beads for the purification of human, mouse & rabbit immunoglobins.
Sigma-Aldrich
Phosphate buffered saline, BioPerformance Certified, pH 7.4
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial