Merck
  • G protein-coupled receptor kinase 2 regulating β2-adrenergic receptor signaling in M2-polarized macrophages contributes to hepatocellular carcinoma progression.
G protein-coupled receptor kinase 2 regulating β2-adrenergic receptor signaling in M2-polarized macrophages contributes to hepatocellular carcinoma progression.
OncoTargets and therapy (2019-08-03)
Jing-Jing Wu, Yang Yang, Wen-Ting Peng, Jia-Chang Sun, Wu-Yi Sun, Wei Wei
ABSTRACT
Background: β2-adrenoceptors (β2-ARs) are expressed on the surface of immune cells, including tumor-associated macrophages (TAMs). Previous studies have demonstrated that the expression of β2-ARs in hepatocellular carcinoma (HCC) is significantly increased in vitro. However, the role of β2-AR in M2-polarized macrophages remains unclear. G protein-coupled receptor kinase 2 (GRK2) can regulate G protein-coupled receptor (GPCR). Previous studies showed that down-regulation of GRK2 in HCC contributes the HCC progression, but it still remains unclear whether the regulation of β2-AR in M2-polarized macrophages by GRK2 can promote HCC. Purpose: The present study was designed to investigate the role of activated β2-AR in M2-polarized macrophages in the HCC progression and GRK2 regulatory effect, as well as the underlying mechanisms involved. Results: The results demonstrated that the M2-polarized macrophages were increased with HCC progression. In vitro, the activation of β2-AR by terbutaline in M2-polarized macrophages elevated the proliferative, migratory and invasive attributes of HCC cells. Furthermore, GRK2 down-regulation in β2-AR activated M2-polarized macrophages activated the downstream cyclic adenosine monophosphate (cAMP)/protein kinase A/cAMP-response element binding protein and cAMP/interleukin-6/signal transducer and the activator of transcription 3 signaling pathways, contributing to the secretion of tumor-associated cytokines, and thus resulting in the promotion of malignant biological behavior in HCC cells. Conclusion: These findings suggest that the regulation of β2-AR occurs through the silencing of GRK2 in M2-polarized macrophages, which is conducive to HCC development, through its engagement in the activation of downstream signaling.
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 40,000