Merck
  • Conditionally fluorescent molecular probes for detecting single base changes in double-stranded DNA.
Conditionally fluorescent molecular probes for detecting single base changes in double-stranded DNA.
Nature chemistry (2013-08-24)
Sherry Xi Chen, David Yu Zhang, Georg Seelig
ABSTRACT
Small variations in nucleic acid sequences can have far-reaching phenotypic consequences. Reliably distinguishing closely related sequences is therefore important for research and clinical applications. Here, we demonstrate that conditionally fluorescent DNA probes are capable of distinguishing variations of a single base in a stretch of target DNA. These probes use a novel programmable mechanism in which each single nucleotide polymorphism generates two thermodynamically destabilizing mismatch bubbles rather than the single mismatch formed during typical hybridization-based assays. Up to a 12,000-fold excess of a target that contains a single nucleotide polymorphism is required to generate the same fluorescence as one equivalent of the intended target, and detection works reliably over a wide range of conditions. Using these probes we detected point mutations in a 198 base-pair subsequence of the Escherichia coli rpoB gene. That our probes are constructed from multiple oligonucleotides circumvents synthesis limitations and enables long continuous DNA sequences to be probed.
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals