Merck
  • Activation of RAGE leads to the release of glutamate from astrocytes and stimulates calcium signal in neurons.
Activation of RAGE leads to the release of glutamate from astrocytes and stimulates calcium signal in neurons.
Journal of cellular physiology (2021-02-12)
Anna Kamynina, Noemi Esteras, Dmitry O Koroev, Plamena R Angelova, Olga M Volpina, Andrey Y Abramov
ABSTRACT
The receptor for advanced glycation end products (RAGE) is a signal receptor first shown to be activated by advanced glycation end products, but also by a variety of signal molecules, including pathological advanced oxidation protein products and β-amyloid. However, most of the RAGE activators have multiple intracellular targets, making it difficult to unravel the exact pathway of RAGE activation. Here, we show that the cell-impermeable RAGE fragment sequence (60-76) of the V-domain of the receptor is able to activate RAGE present on the plasma membrane of neurons and, preferentially, astrocytes. This leads to the exocytosis of vesicular glutamate transporter vesicles and the release of glutamate from astrocytes, which stimulate NMDA and AMPA/kainate receptors, resulting in calcium signals predominantly in neurons. Thus, we show a specific mechanism of RAGE activation by the RAGE fragment and propose a mechanism by which RAGE activation can contribute to the neuronal-astrocytic communication in physiology and pathology.
Sigma-Aldrich
Anti-RAGE Antibody, clone DD/A11 or A11, Chemicon®, from mouse