Merck
  • Deletion of RBMX RGG/RG motif in Shashi-XLID syndrome leads to aberrant p53 activation and neuronal differentiation defects.
Deletion of RBMX RGG/RG motif in Shashi-XLID syndrome leads to aberrant p53 activation and neuronal differentiation defects.
Cell reports (2021-07-15)
Ting Cai, Jessica K Cinkornpumin, Zhenbao Yu, Oscar D Villarreal, William A Pastor, Stéphane Richard
ABSTRACT
RNA-binding proteins play important roles in X-linked intellectual disability (XLID). In this study, we investigate the contribution of the XLID-associated RBMX in neuronal differentiation. We show that RBMX-depleted cells exhibit aberrant activation of the p53 pathway. Moreover, we identify that the RBMX RGG/RG motif is methylated by protein arginine methyltransferase 5 (PRMT5), and this regulates assembly with the SRSF1 splicing factor into higher-order complexes. Depletion of RBMX or disruption of the RBMX/SRSF1 complex in PRMT5-depleted cells reduces SRSF1 binding to the MDM4 precursor (pre-)mRNA, leading to exon 6 exclusion and lower MDM4 protein levels. Transcriptomic analysis of isogenic Shashi-XLID human-induced pluripotent stem cells (hiPSCs) generated using CRISPR-Cas9 reveals a dysregulation of MDM4 splicing and aberrant p53 upregulation. Shashi-XLID neural progenitor cells (NPCs) display differentiation and morphological abnormalities accompanied with excessive apoptosis. Our findings identify RBMX as a regulator of SRSF1 and the p53 pathway, suggesting that the loss of function of the RBMX RGG/RG motif is the cause of Shashi-XLID syndrome.
Sigma-Aldrich
PRMT5/MEP50 Active human, recombinant, expressed in FreeStyle 293-F cells, ≥60% (SDS-PAGE)
Sigma-Aldrich
Poly-L-ornithine solution, mol wt 30,000-70,000, 0.01%, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
GenElute Mammalian Total RNA Miniprep Kit, sufficient for 70 purifications
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture