Merck
  • Galectin-3 Inhibitors Suppress Anoikis Resistance and Invasive Capacity in Thyroid Cancer Cells.
Galectin-3 Inhibitors Suppress Anoikis Resistance and Invasive Capacity in Thyroid Cancer Cells.
International journal of endocrinology (2021-05-27)
Jie-Jen Lee, Yi-Chiung Hsu, Ying-Syuan Li, Shih-Ping Cheng
ABSTRACT
Accumulating evidence suggests that galectin-3 is a histologic marker of thyroid cancer. However, the pharmacological lectin-based approach has not been well studied. In the present study, we aimed to investigate the therapeutic potential of novel galectin-3 inhibitors by treating thyroid cancer cells with different concentrations of GB1107 or TD139. At high doses, TD139, but not GB1107, reduced cell viability and clonogenicity of thyroid cancer cells. TD139 induced apoptosis of thyroid cancer cells, as evident by an increase in the percentage of sub-G1 cells on cell cycle analysis, caspase-3 activation, and PARP1 cleavage. Either GB1107 or TD139 significantly inhibited cell coherence and counteracted anoikis resistance. Both inhibitors decreased migratory and invasive abilities in a dose-dependent manner. Furthermore, GB1107 and TD139 treatment attenuated AKT phosphorylation and decreased the expression of β-catenin and MMP2. In conclusion, these novel galectin-3 inhibitors suppressed the anoikis resistance, motility, and invasive capacity of thyroid cancer cells at least partly through the AKT/β-catenin pathway. Galectin-3 inhibitors are potentially suitable for preclinical evaluation of treatment and/or prevention of metastatic spread in thyroid cancer.
Sigma-Aldrich
Anti-Galectin-3 Antibody, clone M3/38, clone M3/38, from rat
Sigma-Aldrich
Anti-MMP-2 Antibody, a.a. 468-483 hMMP2, clone 42-5D11, clone 42-5D11, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid