Merck
  • Enzyme from the medicinal leech (Hirudo medicinalis) that specifically splits endo-epsilon(-gamma-Glu)-Lys isopeptide bonds: cDNA cloning and protein primary structure.
Enzyme from the medicinal leech (Hirudo medicinalis) that specifically splits endo-epsilon(-gamma-Glu)-Lys isopeptide bonds: cDNA cloning and protein primary structure.
FEBS letters (1996-07-22)
A Fradkov, S Berezhnoy, E Barsova, L Zavalova, S Lukyanov, I Baskova, E D Sverdlov
ABSTRACT
Earlier we detected a novel enzymatic activity in salivary gland secretion of the medicinal leech, splitting isopeptide bonds between the glutamine gamma-carboxamide and lysine epsilon-amino group. This activity is due to destabilase. We described its partial amino acid sequence and sequences to two closely related cDNAs, but none of them perfectly matched the protein isolated. Here we report the isolation and sequence peculiarities of the third cDNA of the family as well as the complete sequence of the destabilase protein. The inferred mature protein product of this cDNA matches the independently determined destabilase protein sequence. It contains 115 amino acid residues including 14 highly conserved Cys residues and is formed from a precursor containing specific leader peptide.