Merck
  • Elucidation of the BMI1 interactome identifies novel regulatory roles in glioblastoma.
Elucidation of the BMI1 interactome identifies novel regulatory roles in glioblastoma.
NAR cancer (2021-07-29)
Verónica Freire-Benéitez, Nicola Pomella, Thomas O Millner, Anaëlle A Dumas, Maria Victoria Niklison-Chirou, Eleni Maniati, Jun Wang, Vinothini Rajeeve, Pedro Cutillas, Silvia Marino
ABSTRACT
Glioblastoma (GBM) is the most common and aggressive intrinsic brain tumour in adults. Epigenetic mechanisms controlling normal brain development are often dysregulated in GBM. Among these, BMI1, a structural component of the Polycomb Repressive Complex 1 (PRC1), which promotes the H2AK119ub catalytic activity of Ring1B, is upregulated in GBM and its tumorigenic role has been shown in vitro and in vivo. Here, we have used protein and chromatin immunoprecipitation followed by mass spectrometry (MS) analysis to elucidate the protein composition of PRC1 in GBM and transcriptional silencing of defining interactors in primary patient-derived GIC lines to assess their functional impact on GBM biology. We identify novel regulatory functions in mRNA splicing and cholesterol transport which could represent novel targetable mechanisms in GBM.
Sigma-Aldrich
Anti-DEDAF Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Rabbit anti-BMI1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
Sigma-Aldrich
Anti-ubiquityl-Histone H2A Antibody, clone E6C5, clone E6C5, Upstate®, from mouse