Merck
  • Androgen-regulated transcription of ESRP2 drives alternative splicing patterns in prostate cancer.
Androgen-regulated transcription of ESRP2 drives alternative splicing patterns in prostate cancer.
eLife (2019-09-04)
Jennifer Munkley, Ling Li, S R Gokul Krishnan, Gerald Hysenaj, Emma Scott, Caroline Dalgliesh, Htoo Zarni Oo, Teresa Mendes Maia, Kathleen Cheung, Ingrid Ehrmann, Karen E Livermore, Hanna Zielinska, Oliver Thompson, Bridget Knight, Paul McCullagh, John McGrath, Malcolm Crundwell, Lorna W Harries, Mads Daugaard, Simon Cockell, Nuno L Barbosa-Morais, Sebastian Oltean, David J Elliott
ABSTRACT
Prostate is the most frequent cancer in men. Prostate cancer progression is driven by androgen steroid hormones, and delayed by androgen deprivation therapy (ADT). Androgens control transcription by stimulating androgen receptor (AR) activity, yet also control pre-mRNA splicing through less clear mechanisms. Here we find androgens regulate splicing through AR-mediated transcriptional control of the epithelial-specific splicing regulator ESRP2. Both ESRP2 and its close paralog ESRP1 are highly expressed in primary prostate cancer. Androgen stimulation induces splicing switches in many endogenous ESRP2-controlled mRNA isoforms, including splicing switches correlating with disease progression. ESRP2 expression in clinical prostate cancer is repressed by ADT, which may thus inadvertently dampen epithelial splice programmes. Supporting this, treatment with the AR antagonist bicalutamide (Casodex) induced mesenchymal splicing patterns of genes including FLNB and CTNND1. Our data reveals a new mechanism of splicing control in prostate cancer with important implications for disease progression.
Sigma-Aldrich
Anti-ESRP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
EZ-Zyme Chromatin Prep Kit, Contains proprietary reagents optimized for the enzymatic shearing of chromatin from mammalian cells at higher resolution than sonication for use in chromatin immunoprecipitation (ChIP) assays.