Merck
  • Quantitative, multiplexed workflow for deep analysis of human blood plasma and biomarker discovery by mass spectrometry.
Quantitative, multiplexed workflow for deep analysis of human blood plasma and biomarker discovery by mass spectrometry.
Nature protocols (2017-07-28)
Hasmik Keshishian, Michael W Burgess, Harrison Specht, Luke Wallace, Karl R Clauser, Michael A Gillette, Steven A Carr
ABSTRACT
Proteomic characterization of blood plasma is of central importance to clinical proteomics and particularly to biomarker discovery studies. The vast dynamic range and high complexity of the plasma proteome have, however, proven to be serious challenges and have often led to unacceptable tradeoffs between depth of coverage and sample throughput. We present an optimized sample-processing pipeline for analysis of the human plasma proteome that provides greatly increased depth of detection, improved quantitative precision and much higher sample analysis throughput as compared with prior methods. The process includes abundant protein depletion, isobaric labeling at the peptide level for multiplexed relative quantification and ultra-high-performance liquid chromatography coupled to accurate-mass, high-resolution tandem mass spectrometry analysis of peptides fractionated off-line by basic pH reversed-phase (bRP) chromatography. The overall reproducibility of the process, including immunoaffinity depletion, is high, with a process replicate coefficient of variation (CV) of <12%. Using isobaric tags for relative and absolute quantitation (iTRAQ) 4-plex, >4,500 proteins are detected and quantified per patient sample on average, with two or more peptides per protein and starting from as little as 200 μl of plasma. The approach can be multiplexed up to 10-plex using tandem mass tags (TMT) reagents, further increasing throughput, albeit with some decrease in the number of proteins quantified. In addition, we provide a rapid protocol for analysis of nonfractionated depleted plasma samples analyzed in 10-plex. This provides ∼600 quantified proteins for each of the ten samples in ∼5 h of instrument time.
Product Description
Millipore
Seppro® Dilution buffer
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Pricing and availability is not currently available.
Millipore
Seppro® Stripping Buffer
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Pricing and availability is not currently available.
Millipore
Seppro® Neutralization Buffer
Pricing and availability is not currently available.