Merck
  • Novel coumarins and 2-thioxo-coumarins as inhibitors of the tumor-associated carbonic anhydrases IX and XII.
Novel coumarins and 2-thioxo-coumarins as inhibitors of the tumor-associated carbonic anhydrases IX and XII.
Bioorganic & medicinal chemistry (2012-03-02)
Fabrizio Carta, Alfonso Maresca, Andrea Scozzafava, Claudiu T Supuran
ABSTRACT
A series of coumarins incorporating tert-butyl-dimethylsilyloxy- or allyoxy- moieties in positions 4-, 6 or 7 of the heterocyclic ring have been synthesized and then converted to the corresponding 2-thioxo-coumarins. Other derivatives incorporating hydroxyethyloxy-, tosylethoxy- and 2-fluroethyloxy- moieties in position 7 of the coumarin ring were synthesized together with derivatives of 4-methyl-7-amino coumarin incorporating acetamido, 3,5-dimethylphenylureido- and tert-butyloxycarbonylamido functionalities. All these compounds were assayed as inhibitors of the metalloenzyme carbonic anhydrase (CA, EC 4.2.1.1). The human (h) cytosolic isoforms hCA I and II were weakly inhibited (hCA I) or not inhibited at all (hCA II) by these (thioxo)coumarins whereas the tumor-associated transmembrane isoforms hCA IX and XII were inhibited with efficiencies from the submicromolar to the low micromolar range by many of these derivatives. The structure-activity relationship for these classes of less investigated CA inhibitors are delineated, with the potential of using them as leads to obtain isoform-selective inhibitors with excellent affinity for CA IX and XII (validated antitumor targets) which do not significantly inhibit the cytosolic offtarget isoforms hCA I and II.
Sigma-Aldrich
Coumarin, ≥99% (HPLC)
Sigma-Aldrich
7-Amino-4-methylcoumarin, 99%
Sigma-Aldrich
7-Amino-4-methylcoumarin, Chromophore for substrates