Merck
A toolbox for the immunomagnetic purification of signaling organelles.
Traffic (Copenhagen, Denmark) (2018-12-21)
Jürgen Fritsch, Vladimir Tchikov, Lena Hennig, Ralph Lucius, Stefan Schütze
ABSTRACT
Homeostasis and the complex functions of organisms and cells rely on the sophisticated spatial and temporal regulation of signaling in different intra- and extracellular compartments and via different mediators. We here present a set of fast and easy to use protocols for the target-specific immunomagnetic enrichment of receptor containing endosomes (receptosomes), plasma membranes, lysosomes and exosomes. Isolation of subcellular organelles and exosomes is prerequisite for and will advance their detailed subsequent biochemical and functional analysis. Sequential application of the different subprotocols allows isolation of morphological and functional intact organelles from one pool of cells. The enrichment is based on a selective labelling using receptor ligands or antibodies together with superparamagnetic microbeads followed by separation in a patented matrix-free high-gradient magnetic purification device. This unique magnetic chamber is based on a focusing system outside of the empty separation column, generating an up to 3 T high-gradient magnetic field focused at the wall of the column.
Sigma-Aldrich
Mouse Anti-Rabbit light chain, HRP conjugate Antibody, Chemicon®, from mouse
Sigma-Aldrich
Goat Anti-Mouse light chain Antibody, HRP conjugate, Chemicon®, from goat