Merck
  • QSAR study and synthesis of new phenyltropanes as ligands of the dopamine transporter (DAT).
QSAR study and synthesis of new phenyltropanes as ligands of the dopamine transporter (DAT).
Bioorganic & medicinal chemistry (2012-02-04)
Sylvie Mavel, Zoya Mincheva, Nathalie Méheux, Yvan Carcenac, Denis Guilloteau, Mohamed Abarbri, Patrick Emond
ABSTRACT
The dopamine transporter (DAT) plays a pivotal role in the regulation of dopamine neurotransmission, and is involved in a number of physiological functions and brain disorders. Furthermore the DAT analysis by molecular imaging techniques is a useful tool for the diagnosis and follow up treatment of diseases involving the DAT. In order to predict the affinity of new derivatives for the DAT, different QSAR molecular modeling models based on cocaine were compared. We have evaluated in these models tropane derivatives synthesized with original synthons which coupled properties of both fluorine and iodine atoms. One compound showed a high in vitro affinity and selectivity for the DAT (K(i)=0.87±0.04 nM). This compound should be radiolabeled with radioiodine for further investigations by SPECT.
Product Description
Supelco
Aniline, analytical standard
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sigma-Aldrich
Naphthalene, 99%
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Sigma-Aldrich
Bromobenzene, ReagentPlus®, 99%
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Supelco
Naphthalene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Bromobenzene, ≥99.5% (GC)
Supelco
Naphthalene, analytical standard
Supelco
Naphthalene solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Biphenyl, ≥99%
Sigma-Aldrich
Benzamide, purified by sublimation, ≥99.5%
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
2-Naphthol, 99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Biphenyl, ReagentPlus®, 99.5%
Sigma-Aldrich
2-Naphthol, 98%
Sigma-Aldrich
Benzamide, 99%
Sigma-Aldrich
2-Naphthol, BioXtra, ≥99.0% (GC)
Supelco
Phenol, PESTANAL®, analytical standard
Supelco
Pyridine, analytical standard
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Supelco
Hexachlorobenzene, PESTANAL®, analytical standard
Supelco
Thymol, analytical standard
Sigma-Aldrich
Phenol, BioUltra, ≥99.5% (GC)
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, BioUltra, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%