Merck
  • Cavin1 intrinsically disordered domains are essential for fuzzy electrostatic interactions and caveola formation.
Cavin1 intrinsically disordered domains are essential for fuzzy electrostatic interactions and caveola formation.
Nature communications (2021-02-12)
Vikas A Tillu, James Rae, Ya Gao, Nicholas Ariotti, Matthias Floetenmeyer, Oleksiy Kovtun, Kerrie-Ann McMahon, Natasha Chaudhary, Robert G Parton, Brett M Collins
ABSTRACT
Caveolae are spherically shaped nanodomains of the plasma membrane, generated by cooperative assembly of caveolin and cavin proteins. Cavins are cytosolic peripheral membrane proteins with negatively charged intrinsically disordered regions that flank positively charged α-helical regions. Here, we show that the three disordered domains of Cavin1 are essential for caveola formation and dynamic trafficking of caveolae. Electrostatic interactions between disordered regions and α-helical regions promote liquid-liquid phase separation behaviour of Cavin1 in vitro, assembly of Cavin1 oligomers in solution, generation of membrane curvature, association with caveolin-1, and Cavin1 recruitment to caveolae in cells. Removal of the first disordered region causes irreversible gel formation in vitro and results in aberrant caveola trafficking through the endosomal system. We propose a model for caveola assembly whereby fuzzy electrostatic interactions between Cavin1 and caveolin-1 proteins, combined with membrane lipid interactions, are required to generate membrane curvature and a metastable caveola coat.
Sigma-Aldrich
D-(+)-Maltose monohydrate, Type II, ≥95%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Cholesterol-Water Soluble, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
3,3′-Diaminobenzidine tetrahydrochloride, tablet, 10 mg substrate per tablet
Sigma-Aldrich
Polyethylenimine, branched, average Mw ~25,000 by LS, average Mn ~10,000 by GPC, branched
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)
Sigma-Aldrich
Benzamidine hydrochloride hydrate, ≥99%