Merck
Purification of high-quality micro RNA from the heart tissue.
Acta biologica Hungarica (2011-11-29)
Nóra Faragó, Agnes Zvara, Z Varga, P Ferdinandy, L G Puskás
ABSTRACT
Micro RNAs (miRNA) are an abundant class of small RNAs that regulate the stability and translation of cognate mRNAs. MiRNAs are potential diagnostic markers, moreover, they play an essential role in the development of various heart disesases. In case of limited tissue material, such as, e.g. human biopsies, purification of miRNAs with sufficient yield is critical. Reproducible expression analysis of miRNAs is highly dependent on the quality of the RNA, which is often difficult to achieve from fibrous tissue such as the heart. Several companies developed general purification kits for miRNAs, however, none of them are specialized to fibrotic tissues. Here we describe an optimized miRNA purification protocol that results in high miRNA yield as compared to other methods including trizol-based and column-based protocols. By using our improved protocol, miRNA obtained from heart tissue gave more reproducible results in QRT-PCR analysis and obtained more significant calls (172 vs. 118) during DNA microarray analysis when compared to the commercially available kit. In addition to the heart tissue, the present protocol can be applied to other fibrotic tissues, such as lung or skeletal muscle to isolate high-purity miRNA.
Product Description
Roche
High Pure miRNA Isolation Kit, kit of for 50 isolations