Merck
Detection of NDM-1-producing Klebsiella pneumoniae in Kenya.
Antimicrobial agents and chemotherapy (2010-12-01)
Laurent Poirel, Gunturu Revathi, Sandrine Bernabeu, Patrice Nordmann
ABSTRACT
Seven carbapenem-resistant NDM-1-positive Klebsiella pneumoniae isolates were recovered from patients hospitalized between 2007 and 2009 in different wards at a referral and tertiary care center in Nairobi. Most of the isolates were obtained from urine. All isolates carried the bla(NDM-1) carbapenemase gene previously reported from India, Pakistan, and the United Kingdom. These isolates were clonally related and expressed many other resistance determinants, including β-lactamases CTX-M-15, OXA-1, OXA-9, CMY-6, and aminoglycoside resistance methylase RmtC. This work corresponds to the first report of NDM-1 producers in Africa.
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Nitrofurantoin, 98.0-102.0% (EP, UV)
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Supelco
Nitrofurantoin, VETRANAL®, analytical standard