Merck
  • Oxidized Alginate-Based Hydrogels for Tissue Engineering Applications: A Review.
Oxidized Alginate-Based Hydrogels for Tissue Engineering Applications: A Review.
Biomacromolecules (2017-11-28)
Supachai Reakasame, Aldo R Boccaccini
ABSTRACT
Oxidized alginate (OA)-based hydrogels have drawn considerable attention as biodegradable materials for tissue engineering applications. OA possesses a faster degradation rate and contains more reactive groups compared to native alginate. This review summarizes the research publications reporting the development of OA-based hydrogels for tissue engineering applications including bone, cartilage, blood vessel, cornea, and other soft tissues, highlighting OA key properties and processing approaches.
Sigma-Aldrich
Alginate Aldehyde,
20% aldehyde content, medium viscosity
Sigma-Aldrich
Alginate Aldehyde, 35% aldehyde content, medium viscosity